Product Details

SNP ID
rs1045389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39539365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCTCTTGAAGTAGTGTCACCAC[A/C]TAACTATTCTCTATTGATAAACTTA
Phenotype
MIM: 609773
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EID2 PubMed Links

Gene Details

Gene
EID2
Gene Name
EP300 interacting inhibitor of differentiation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153232.3 834 UTR 3 NP_694964.3
Gene
EID2B
Gene Name
EP300 interacting inhibitor of differentiation 2B
There are no transcripts associated with this gene.

View Full Product Details