Product Details

SNP ID
rs1048552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45162976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTGAGCAGCTACTGGGCAGTGAC[G/T]CTGCCGAGGCGGGAATCCCACCACA
Phenotype
MIM: 610396
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NKPD1 PubMed Links

Gene Details

Gene
NKPD1
Gene Name
NTPase, KAP family P-loop domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198478.3 721 Intron NP_940880.3
XM_011526799.2 721 Intron XP_011525101.1
XM_011526802.2 721 Intron XP_011525104.1
XM_011526804.2 721 Intron XP_011525106.1
XM_011526805.2 721 Intron XP_011525107.1
Gene
TRAPPC6A
Gene Name
trafficking protein particle complex 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270891.1 721 UTR 3 NP_001257820.1
NM_001270892.1 721 UTR 3 NP_001257821.1
NM_001270893.1 721 UTR 3 NP_001257822.1
NM_024108.2 721 UTR 3 NP_077013.1
XM_017027295.1 721 UTR 3 XP_016882784.1

View Full Product Details