Product Details

SNP ID
rs1056852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41264582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCTGGACACTCGAGGCCTCAAG[A/G]CCGAGCTTGCTGAGCGGCTGCAGGC
Phenotype
MIM: 109135 MIM: 605800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AXL PubMed Links

Gene Details

Gene
AXL
Gene Name
AXL receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
HNRNPUL1
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301016.2 409 Intron NP_001287945.1
NM_001321208.1 409 UTR 5 NP_001308137.1
NM_001321211.1 409 Intron NP_001308140.1
NM_007040.5 409 Missense Mutation ACC,GCC T27A NP_008971.2
NM_144732.4 409 Intron NP_653333.1
XM_005258459.3 409 Missense Mutation ACC,GCC T27A XP_005258516.1
XM_005258461.4 409 Intron XP_005258518.1
XM_005258463.3 409 UTR 5 XP_005258520.1
XM_005258464.3 409 Intron XP_005258521.1
XM_005258465.4 409 Intron XP_005258522.1
XM_011526392.2 409 Missense Mutation ACC,GCC T27A XP_011524694.1
XM_011526393.2 409 Intron XP_011524695.1
XM_011526394.2 409 Intron XP_011524696.1
XM_011526395.2 409 Intron XP_011524697.1

View Full Product Details