Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127384.2 | 3925 | UTR 3 | NP_001120856.1 | ||
NM_001291133.1 | 3925 | Intron | NP_001278062.1 | ||
NM_013266.3 | 3925 | UTR 3 | NP_037398.2 | ||
XM_017016151.1 | 3925 | UTR 3 | XP_016871640.1 | ||
XM_017016152.1 | 3925 | UTR 3 | XP_016871641.1 | ||
XM_017016153.1 | 3925 | UTR 3 | XP_016871642.1 | ||
XM_017016154.1 | 3925 | UTR 3 | XP_016871643.1 | ||
XM_017016155.1 | 3925 | UTR 3 | XP_016871644.1 | ||
XM_017016156.1 | 3925 | UTR 3 | XP_016871645.1 | ||
XM_017016157.1 | 3925 | UTR 3 | XP_016871646.1 | ||
XM_017016158.1 | 3925 | Intron | XP_016871647.1 |