Product Details

SNP ID
rs8413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136428859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATCCTGAACTTCCAGCAGTTCCC[C/T]AGCCCGGTTCCACTAATGTCATTCC
Phenotype
MIM: 613037 MIM: 613036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INPP5E PubMed Links

Gene Details

Gene
INPP5E
Gene Name
inositol polyphosphate-5-phosphatase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318502.1 3146 UTR 3 NP_001305431.1
NM_019892.5 3146 UTR 3 NP_063945.2
XM_017014926.1 3146 UTR 3 XP_016870415.1
Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
There are no transcripts associated with this gene.

View Full Product Details