Product Details
- SNP ID
-
rs954504
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37554492 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATCTTGTTACTTTTGTCCCCATGT[A/G]TGTATCTTTTTTCCATCAGGCTGTT
- Phenotype
-
MIM: 613903
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF540
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114609667] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF540
- Gene Name
- zinc finger protein 540
- Gene
- ZNF571
- Gene Name
- zinc finger protein 571
There are no transcripts associated with this gene.
- Gene
- ZNF571-AS1
- Gene Name
- ZNF571 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details