Product Details

SNP ID
rs1548803
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:10806432 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAATTGCAAACAGGTTAAAGAGAGT[C/T]AAGGGTTCAAAGTATGGTATTTTAC
Phenotype
MIM: 604793 MIM: 604794 MIM: 604795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R7 PubMed Links

Gene Details

Gene
TAS2R7
Gene Name
taste 2 receptor member 7
There are no transcripts associated with this gene.

Gene
TAS2R8
Gene Name
taste 2 receptor member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023918.1 549 Silent Mutation TTA,TTG L183L NP_076407.1
Gene
TAS2R9
Gene Name
taste 2 receptor member 9
There are no transcripts associated with this gene.

View Full Product Details