Product Details

SNP ID
rs1053876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38878797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTAGCCACAGGCCCCCGGGGG[A/C]AGGAGACAGAGTGGGGGCCCGAAGC
Phenotype
MIM: 604480
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195833.1 1715 Intron NP_001182762.1
NM_198445.3 1715 Intron NP_940847.1
XM_006723015.2 1715 Intron XP_006723078.1
XM_006723016.2 1715 Intron XP_006723079.1
XM_011526454.2 1715 Intron XP_011524756.1
XM_011526455.2 1715 Intron XP_011524757.1
Gene
SIRT2
Gene Name
sirtuin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193286.1 1715 UTR 3 NP_001180215.1
NM_012237.3 1715 UTR 3 NP_036369.2
NM_030593.2 1715 UTR 3 NP_085096.1
XM_006723111.1 1715 UTR 3 XP_006723174.1
XM_011526654.1 1715 UTR 3 XP_011524956.1
XM_011526655.1 1715 UTR 3 XP_011524957.1
XM_017026500.1 1715 UTR 3 XP_016881989.1

View Full Product Details