Product Details

SNP ID
rs11209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5386260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTTGCTTCACCATTTCCCTTAT[A/G]TGTTCACCCCTGTAAAATTGTAAAG
Phenotype
MIM: 602552 MIM: 603616
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP88 PubMed Links

Gene Details

Gene
NUP88
Gene Name
nucleoporin 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320653.1 2359 Silent Mutation CAC,CAT H740H NP_001307582.1
NM_002532.5 2359 Silent Mutation CAC,CAT H724H NP_002523.2
XM_011523893.2 2359 Silent Mutation CAC,CAT H609H XP_011522195.1
XM_017024694.1 2359 Silent Mutation CAC,CAT H593H XP_016880183.1
Gene
RABEP1
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083585.2 2359 UTR 3 NP_001077054.1
NM_001291581.1 2359 UTR 3 NP_001278510.1
NM_001291582.1 2359 Intron NP_001278511.1
NM_004703.5 2359 UTR 3 NP_004694.2
XM_011524060.1 2359 Intron XP_011522362.1
XM_017025294.1 2359 Intron XP_016880783.1
XM_017025295.1 2359 Intron XP_016880784.1

View Full Product Details