Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320653.1 | 2344 | Silent Mutation | NP_001307582.1 | ||
NM_002532.5 | 2344 | Silent Mutation | NP_002523.2 | ||
XM_011523893.2 | 2344 | Silent Mutation | XP_011522195.1 | ||
XM_017024694.1 | 2344 | Silent Mutation | XP_016880183.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001083585.2 | 2344 | Intron | NP_001077054.1 | ||
NM_001291581.1 | 2344 | Intron | NP_001278510.1 | ||
NM_001291582.1 | 2344 | Intron | NP_001278511.1 | ||
NM_004703.5 | 2344 | Intron | NP_004694.2 | ||
XM_011524060.1 | 2344 | Intron | XP_011522362.1 | ||
XM_017025294.1 | 2344 | Intron | XP_016880783.1 | ||
XM_017025295.1 | 2344 | Intron | XP_016880784.1 |