Product Details

SNP ID
rs1182968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68540042 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGAGAGGGCCAGTGTGGTGCGAT[C/T]TTGTGGGAGGAGTTGAGATTGGATG
Phenotype
MIM: 609451
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP90 PubMed Links

Gene Details

Gene
ZFP90
Gene Name
ZFP90 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305203.1 Intron NP_001292132.1
NM_001305204.1 Intron NP_001292133.1
NM_001305206.1 Intron NP_001292135.1
NM_001305207.1 Intron NP_001292136.1
NM_001305208.1 Intron NP_001292137.1
NM_133458.3 Intron NP_597715.2
XM_005255804.2 Intron XP_005255861.1
XM_017022952.1 Intron XP_016878441.1

View Full Product Details