Product Details
- SNP ID
-
rs1128868
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:43556737 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTTTCTTGTGAACCCGCTGGTGG[A/C]TGTGAAGGTTGGAGCTCTGGCTGAA
- Phenotype
-
MIM: 601069
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF239
PubMed Links
Gene Details
- Gene
- ZNF239
- Gene Name
- zinc finger protein 239
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001099282.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001092752.1 |
NM_001099283.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001092753.1 |
NM_001099284.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001092754.1 |
NM_001324347.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001311276.1 |
NM_001324348.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001311277.1 |
NM_001324349.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001311278.1 |
NM_001324350.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001311279.1 |
NM_001324351.1 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_001311280.1 |
NM_001324352.1 |
1514 |
Missense Mutation |
AGC,ATC |
S490I |
NP_001311281.1 |
NM_001324353.1 |
1514 |
Missense Mutation |
AGC,ATC |
S561I |
NP_001311282.1 |
NM_005674.2 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
NP_005665.2 |
XM_005271832.2 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
XP_005271889.1 |
XM_006718001.2 |
1514 |
Missense Mutation |
AGC,ATC |
S490I |
XP_006718064.1 |
XM_006718003.3 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
XP_006718066.1 |
XM_011540232.2 |
1514 |
Missense Mutation |
AGC,ATC |
S592I |
XP_011538534.1 |
XM_011540234.2 |
1514 |
Missense Mutation |
AGC,ATC |
S490I |
XP_011538536.1 |
XM_011540235.2 |
1514 |
Missense Mutation |
AGC,ATC |
S490I |
XP_011538537.1 |
XM_011540236.2 |
1514 |
Missense Mutation |
AGC,ATC |
S490I |
XP_011538538.1 |
XM_011540237.2 |
1514 |
Missense Mutation |
AGC,ATC |
S490I |
XP_011538539.1 |
XM_011540238.2 |
1514 |
Missense Mutation |
AGC,ATC |
S448I |
XP_011538540.1 |
XM_017016740.1 |
1514 |
Missense Mutation |
AGC,ATC |
S490I |
XP_016872229.1 |
View Full Product Details