Product Details

SNP ID
rs1421139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94176580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACAGATTAACATTTAAATTTTT[C/T]ACATATGGTATATTTCAGAAATTTG
Phenotype
MIM: 605386 MIM: 612145 MIM: 612146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIRLET7A1 PubMed Links

Gene Details

Gene
MIRLET7A1
Gene Name
microRNA let-7a-1
There are no transcripts associated with this gene.

Gene
MIRLET7D
Gene Name
microRNA let-7d
There are no transcripts associated with this gene.

Gene
MIRLET7DHG
Gene Name
MIRLET7D host gene
There are no transcripts associated with this gene.

Gene
MIRLET7F1
Gene Name
microRNA let-7f-1
There are no transcripts associated with this gene.

View Full Product Details