Product Details

SNP ID
rs1192793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101281409 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGATGACATCTCTACGTGGGGGA[C/T]GACTATTTTCATTTTTTCCTTATGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF149 PubMed Links

Gene Details

Gene
RNF149
Gene Name
ring finger protein 149
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173647.3 1408 Intron NP_775918.2
XM_005263920.2 1408 UTR 3 XP_005263977.1
XM_005263921.3 1408 Intron XP_005263978.1
XM_011510990.2 1408 UTR 3 XP_011509292.1
Gene
SNORD89
Gene Name
small nucleolar RNA, C/D box 89
There are no transcripts associated with this gene.

View Full Product Details