Product Details
- SNP ID
-
rs3900863
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:49948746 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGTATATTTAAATATCATTAATC[A/G]TCATAATACTACAGATAGACTTAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB133
PubMed Links
Gene Details
- Gene
- DEFB133
- Gene Name
- defensin beta 133
View Full Product Details