Product Details

SNP ID
rs1041320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32390939 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGGCGAAAGACCTGAAAAAATA[A/C]AACTGAATAATATTGGGTCACTTGT
Phenotype
MIM: 100880
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACO1 PubMed Links

Gene Details

Gene
ACO1
Gene Name
aconitase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278352.1 Intron NP_001265281.1
NM_002197.2 Intron NP_002188.1
XM_005251476.1 Intron XP_005251533.1
XM_011517888.1 Intron XP_011516190.1

View Full Product Details