Product Details

SNP ID
rs704581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:99837316 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTTAGTTAGCTCCTGTTTATAT[A/T]AAACAGTGGTTTTCTGAGGCTTCAA
Phenotype
MIM: 612993
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CMSS1 PubMed Links

Gene Details

Gene
CMSS1
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167924.1 Intron NP_001161396.1
NM_032359.3 Intron NP_115735.2
Gene
FILIP1L
Gene Name
filamin A interacting protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042459.2 Intron NP_001035924.1
NM_001282793.1 Intron NP_001269722.1
NM_001282794.1 Intron NP_001269723.1
NM_014890.3 Intron NP_055705.2
NM_182909.3 Intron NP_878913.2
XM_006713486.2 Intron XP_006713549.1
XM_011512371.2 Intron XP_011510673.1

View Full Product Details