Product Details

SNP ID
rs816411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56103796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGTTTCAAGCACACCGGAAATA[C/T]AAGAGTATACACACCTATGCTCATT
Phenotype
MIM: 616244
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD2 PubMed Links

Gene Details

Gene
CHCHD2
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320327.1 Intron NP_001307256.1
NM_016139.3 Intron NP_057223.1

View Full Product Details