Product Details

SNP ID
rs816406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56106478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCGAAGAGCTAAGCGACTTCTG[A/C]GGAGACCGGAAGATGGGAGGCGGGG
Phenotype
MIM: 616244
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHCHD2 PubMed Links

Gene Details

Gene
CHCHD2
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320327.1 Intron NP_001307256.1
NM_016139.3 Intron NP_057223.1
Gene
NUPR2
Gene Name
nuclear protein 2, transcriptional regulator
There are no transcripts associated with this gene.

View Full Product Details