Product Details

SNP ID
rs914842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120856693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAGGGCTTCCCTTTGAGGGAACC[A/G]AATGATGCAGGCTCTTTAAAAATTT
Phenotype
MIM: 609740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 3118 Intron NP_001009936.1
NM_001286840.1 3118 UTR 3 NP_001273769.1
NM_001286842.1 3118 UTR 3 NP_001273771.1
NM_001286843.1 3118 Intron NP_001273772.1
NM_015651.2 3118 UTR 3 NP_056466.1
XM_005251906.2 3118 UTR 3 XP_005251963.1
XM_011518509.2 3118 UTR 3 XP_011516811.1
XM_011518511.2 3118 Intron XP_011516813.1
XM_011518515.2 3118 Intron XP_011516817.1
XM_011518516.2 3118 Intron XP_011516818.1
XM_017014612.1 3118 UTR 3 XP_016870101.1
XM_017014613.1 3118 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details