Product Details

SNP ID
rs873836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:126805336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGCAGTCTTCTGCCCACACGGCG[C/T]GCAGACCGCGATACCGAGGCCTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB43 PubMed Links

Gene Details

Gene
ZBTB43
Gene Name
zinc finger and BTB domain containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135776.1 214 Intron NP_001129248.1
NM_014007.3 214 Intron NP_054726.1
XM_005251835.3 214 UTR 5 XP_005251892.1
XM_011518409.1 214 Intron XP_011516711.1
XM_011518411.2 214 Intron XP_011516713.1

View Full Product Details