Product Details
- SNP ID
-
rs942164
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:98122752 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCCCTACCTCCTGGGGGACTCTTC[C/T]GTTAACTAGGGCTGAAATCTGCCCA
- Phenotype
-
MIM: 602159
MIM: 606556
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CORO2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147680752] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CORO2A
- Gene Name
- coronin 2A
- Gene
- TRIM14
- Gene Name
- tripartite motif containing 14
There are no transcripts associated with this gene.
View Full Product Details