Product Details

SNP ID
rs876577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:471158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCACCTGCCCTGGATCTGATCATC[A/C]ATTATATCATCTCTTCCATCTGATG
Phenotype
MIM: 611432 MIM: 607704
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DOCK8 PubMed Links

Gene Details

Gene
DOCK8
Gene Name
dedicator of cytokinesis 8
There are no transcripts associated with this gene.

Gene
KANK1
Gene Name
KN motif and ankyrin repeat domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256876.1 Intron NP_001243805.1
NM_001256877.1 Intron NP_001243806.1
NM_015158.3 Intron NP_055973.2
NM_153186.4 Intron NP_694856.1
XM_005251411.2 Intron XP_005251468.1
XM_005251414.3 Intron XP_005251471.1
XM_005251415.2 Intron XP_005251472.1
XM_005251416.2 Intron XP_005251473.1
XM_005251417.3 Intron XP_005251474.1
XM_005251418.2 Intron XP_005251475.1
XM_005251419.2 Intron XP_005251476.1
XM_006716743.2 Intron XP_006716806.1
XM_011517819.1 Intron XP_011516121.1
XM_011517820.1 Intron XP_011516122.1
XM_011517821.1 Intron XP_011516123.1
XM_017014511.1 Intron XP_016870000.1
XM_017014512.1 Intron XP_016870001.1
XM_017014513.1 Intron XP_016870002.1
XM_017014514.1 Intron XP_016870003.1
XM_017014515.1 Intron XP_016870004.1
XM_017014516.1 Intron XP_016870005.1
XM_017014517.1 Intron XP_016870006.1
XM_017014518.1 Intron XP_016870007.1
XM_017014519.1 Intron XP_016870008.1
XM_017014520.1 Intron XP_016870009.1
XM_017014521.1 Intron XP_016870010.1
XM_017014522.1 Intron XP_016870011.1
XM_017014523.1 Intron XP_016870012.1
XM_017014524.1 Intron XP_016870013.1
XM_017014525.1 Intron XP_016870014.1
XM_017014526.1 Intron XP_016870015.1
XM_017014527.1 Intron XP_016870016.1
XM_017014528.1 Intron XP_016870017.1
XM_017014529.1 Intron XP_016870018.1
XM_017014530.1 Intron XP_016870019.1
XM_017014531.1 Intron XP_016870020.1
XM_017014532.1 Intron XP_016870021.1
XM_017014533.1 Intron XP_016870022.1
XM_017014534.1 Intron XP_016870023.1
XM_017014535.1 Intron XP_016870024.1
XM_017014536.1 Intron XP_016870025.1
XM_017014537.1 Intron XP_016870026.1
XM_017014538.1 Intron XP_016870027.1
XM_017014539.1 Intron XP_016870028.1
XM_017014540.1 Intron XP_016870029.1
XM_017014541.1 Intron XP_016870030.1
XM_017014542.1 Intron XP_016870031.1

View Full Product Details