Product Details

SNP ID
rs1547489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45431932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGAATTTTCAATCAATTTTAGTT[A/G]CTACTGATGGTTCCTGATCATTAGC
Phenotype
MIM: 608409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf48 PubMed Links

Gene Details

Gene
C15orf48
Gene Name
chromosome 15 open reading frame 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032413.3 Intron NP_115789.1
NM_197955.2 Intron NP_922946.1
Gene
MIR147B
Gene Name
microRNA 147b
There are no transcripts associated with this gene.

View Full Product Details