Product Details

SNP ID
rs1693531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52560413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGCTGCAAATGTCATTTATATT[C/T]AGAATGAATTTATAACATGTCTTAG
Phenotype
MIM: 605487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPP19 PubMed Links

Gene Details

Gene
ARPP19
Gene Name
cAMP regulated phosphoprotein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306191.1 Intron NP_001293120.1
NM_001306195.1 Intron NP_001293124.1
NM_001306196.1 Intron NP_001293125.1
NM_006628.5 Intron NP_006619.1
XM_017021868.1 Intron XP_016877357.1
XM_017021869.1 Intron XP_016877358.1
XM_017021870.1 Intron XP_016877359.1
XM_017021871.1 Intron XP_016877360.1

View Full Product Details