Product Details

SNP ID
rs607594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45642642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACTGCCCTCCTTTATATCCACC[A/G]TTAGGAGATTCTCATTGCAAGTGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SQRDL PubMed Links

Gene Details

Gene
SQRDL
Gene Name
sulfide quinone reductase-like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271213.1 Intron NP_001258142.1
NM_021199.3 Intron NP_067022.1

View Full Product Details