Product Details

SNP ID
rs1802452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49129506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATAAAAGGAATATGTATTCTTG[A/T]GTAAGGCTTAATTAATTTTATAAGC
Phenotype
MIM: 604508
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COPS2 PubMed Links

Gene Details

Gene
COPS2
Gene Name
COP9 signalosome subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143887.1 1199 Missense Mutation ACA,TCA T374S NP_001137359.1
NM_004236.3 1199 Missense Mutation ACA,TCA T367S NP_004227.1

View Full Product Details