Product Details

SNP ID
rs1043156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106978499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATTTCTTCAACATAGGTTTCATC[C/T]TCTAAAAGTACCCATCCTTTTAATC
Phenotype
MIM: 616929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTERF2 PubMed Links

Gene Details

Gene
MTERF2
Gene Name
mitochondrial transcription termination factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033050.2 572 Silent Mutation GAA,GAG E72E NP_001028222.1
NM_025198.4 572 Silent Mutation GAA,GAG E72E NP_079474.2
XM_011538767.2 572 Silent Mutation GAA,GAG E72E XP_011537069.1
XM_017019985.1 572 Silent Mutation GAA,GAG E72E XP_016875474.1
XM_017019986.1 572 Silent Mutation GAA,GAG E72E XP_016875475.1
XM_017019987.1 572 Silent Mutation GAA,GAG E72E XP_016875476.1
Gene
TMEM263
Gene Name
transmembrane protein 263
There are no transcripts associated with this gene.

View Full Product Details