Product Details

SNP ID
rs1565533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44702740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGTCATTAAAAAATGAAATATA[C/T]GCCACTGGAGTCATATCACAAATGG
Phenotype
MIM: 613222
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNPDA2 PubMed Links

Gene Details

Gene
GNPDA2
Gene Name
glucosamine-6-phosphate deaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270880.1 1266 UTR 3 NP_001257809.1
NM_001270881.1 1266 UTR 3 NP_001257810.1
NM_138335.2 1266 UTR 3 NP_612208.1
Gene
GUF1
Gene Name
GUF1 homolog, GTPase
There are no transcripts associated with this gene.

View Full Product Details