Product Details

SNP ID
rs138057
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.22:43825367 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
CCGCCCGCTGGAAGTCTCTGGCAGA[A/G]GGACGTCCATTCTGAAGACCTAGTG
Phenotype
MIM: 608359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULT4A1 PubMed Links

Gene Details

Gene
SULT4A1
Gene Name
sulfotransferase family 4A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014351.3 1210 UTR 3 NP_055166.1
XM_011530120.2 1210 Intron XP_011528422.1
XM_011530121.1 1210 UTR 3 XP_011528423.1

View Full Product Details