Product Details
- SNP ID
-
rs1199473
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:70270712 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGCTAGATGTGACTTTCAGGCTG[C/T]CATTTTGTAACCTCTGGTATACGTC
- Phenotype
-
MIM: 301770
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARR3
PubMed Links
Gene Details
- Gene
- ARR3
- Gene Name
- arrestin 3
View Full Product Details