Product Details

SNP ID
rs1049863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:60029424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATACCCTGACAGTTTGGGTCCAA[A/G]AAAAATAAGGCGAGCTGTTGTAGAT
Phenotype
MIM: 600465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANK3 PubMed Links

Gene Details

Gene
ANK3
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001149.3 8513 UTR 3 NP_001140.2
NM_001204403.1 8513 UTR 3 NP_001191332.1
NM_001204404.1 8513 UTR 3 NP_001191333.1
NM_001320874.1 8513 UTR 3 NP_001307803.1
NM_020987.4 8513 UTR 3 NP_066267.2
XM_005269715.3 8513 UTR 3 XP_005269772.1
XM_006717796.3 8513 UTR 3 XP_006717859.1
XM_006717802.3 8513 UTR 3 XP_006717865.1
XM_011539708.2 8513 UTR 3 XP_011538010.1
XM_011539709.2 8513 UTR 3 XP_011538011.1
XM_017016110.1 8513 UTR 3 XP_016871599.1
XM_017016111.1 8513 UTR 3 XP_016871600.1
XM_017016112.1 8513 UTR 3 XP_016871601.1
XM_017016113.1 8513 UTR 3 XP_016871602.1
XM_017016114.1 8513 UTR 3 XP_016871603.1
XM_017016115.1 8513 UTR 3 XP_016871604.1
XM_017016116.1 8513 UTR 3 XP_016871605.1
XM_017016117.1 8513 UTR 3 XP_016871606.1
XM_017016118.1 8513 UTR 3 XP_016871607.1
XM_017016119.1 8513 UTR 3 XP_016871608.1
XM_017016120.1 8513 UTR 3 XP_016871609.1
XM_017016121.1 8513 UTR 3 XP_016871610.1
XM_017016122.1 8513 UTR 3 XP_016871611.1
XM_017016123.1 8513 UTR 3 XP_016871612.1
XM_017016124.1 8513 UTR 3 XP_016871613.1
XM_017016125.1 8513 UTR 3 XP_016871614.1
XM_017016126.1 8513 UTR 3 XP_016871615.1
XM_017016127.1 8513 UTR 3 XP_016871616.1
XM_017016128.1 8513 UTR 3 XP_016871617.1
XM_017016129.1 8513 UTR 3 XP_016871618.1
XM_017016130.1 8513 UTR 3 XP_016871619.1
XM_017016131.1 8513 UTR 3 XP_016871620.1
XM_017016132.1 8513 UTR 3 XP_016871621.1
XM_017016133.1 8513 UTR 3 XP_016871622.1
XM_017016134.1 8513 UTR 3 XP_016871623.1
XM_017016135.1 8513 UTR 3 XP_016871624.1
XM_017016136.1 8513 UTR 3 XP_016871625.1
XM_017016137.1 8513 UTR 3 XP_016871626.1
XM_017016138.1 8513 UTR 3 XP_016871627.1
XM_017016139.1 8513 UTR 3 XP_016871628.1
XM_017016140.1 8513 UTR 3 XP_016871629.1
XM_017016141.1 8513 UTR 3 XP_016871630.1
XM_017016142.1 8513 UTR 3 XP_016871631.1
XM_017016143.1 8513 UTR 3 XP_016871632.1
XM_017016144.1 8513 UTR 3 XP_016871633.1
XM_017016145.1 8513 UTR 3 XP_016871634.1
XM_017016146.1 8513 UTR 3 XP_016871635.1
XM_017016147.1 8513 UTR 3 XP_016871636.1
XM_017016148.1 8513 UTR 3 XP_016871637.1
XM_017016149.1 8513 UTR 3 XP_016871638.1
XM_017016150.1 8513 UTR 3 XP_016871639.1

View Full Product Details