Product Details

SNP ID
rs1043554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127573139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACACTTCAGTCCATCCACCATGG[T/G]GGATGCTTTGAGGAGGCCAAAGCAC
Phenotype
MIM: 608336
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6825 PubMed Links

Gene Details

Gene
MIR6825
Gene Name
microRNA 6825
There are no transcripts associated with this gene.

Gene
TPRA1
Gene Name
transmembrane protein adipocyte associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136053.2 1905 UTR 3 NP_001129525.1
NM_001142646.2 1905 UTR 3 NP_001136118.1
XM_006713495.3 1905 UTR 3 XP_006713558.1
XM_006713496.2 1905 UTR 3 XP_006713559.1
XM_006713497.3 1905 UTR 3 XP_006713560.1
XM_011512422.1 1905 UTR 3 XP_011510724.1
XM_017005707.1 1905 UTR 3 XP_016861196.1
XM_017005708.1 1905 UTR 3 XP_016861197.1
XM_017005709.1 1905 UTR 3 XP_016861198.1
XM_017005710.1 1905 UTR 3 XP_016861199.1

View Full Product Details