Product Details

SNP ID
rs1059333
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:6461546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGCGGGCTGCTGCTGGCGCCAG[C/T]GCTTGAGCATCTCGTACTGCTGGTC
Phenotype
MIM: 606351 MIM: 611101 MIM: 603366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031475.2 1119 Intron NP_113663.2
XM_005263501.2 1119 Intron XP_005263558.1
XM_011542231.1 1119 Intron XP_011540533.1
XM_011542232.1 1119 Intron XP_011540534.1
XM_011542233.2 1119 Intron XP_011540535.1
XM_011542236.2 1119 Intron XP_011540538.1
XM_011542237.1 1119 Intron XP_011540539.1
XM_011542238.2 1119 Intron XP_011540540.1
XM_017002433.1 1119 Intron XP_016857922.1
XM_017002434.1 1119 Intron XP_016857923.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

Gene
TNFRSF25
Gene Name
TNF receptor superfamily member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039664.1 1119 Intron NP_001034753.1
NM_003790.2 1119 Missense Mutation CAC,CGC H381R NP_003781.1
NM_148965.1 1119 Missense Mutation CAC,CGC H390R NP_683866.1
NM_148966.1 1119 Missense Mutation CAC,CGC H344R NP_683867.1
NM_148967.1 1119 Missense Mutation CAC,CGC H336R NP_683868.1
NM_148970.1 1119 Missense Mutation CAC,CGC H198R NP_683871.1

View Full Product Details