Product Details

SNP ID
rs1045854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48644489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGTCATAAAGAACCCTCTTCA[C/G]CTCCACATCCACGGGGCTCTGCCGC
Phenotype
MIM: 604644 MIM: 124097
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CA11 PubMed Links

Gene Details

Gene
CA11
Gene Name
carbonic anhydrase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001217.4 903 Missense Mutation CTG,GTG L75V NP_001208.2
Gene
DBP
Gene Name
D-box binding PAR bZIP transcription factor
There are no transcripts associated with this gene.

Gene
SEC1P
Gene Name
secretory blood group 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details