Product Details
- SNP ID
-
rs1480274
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:121668495 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCATCTTCTCCACAGAGCAGCAG[A/C]AGAGCTTTCTTATAGTCCCCAGATG
- Phenotype
-
MIM: 131230
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA5
PubMed Links
Gene Details
- Gene
- ANXA5
- Gene Name
- annexin A5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001154.3 |
1101 |
Silent Mutation |
CTG,CTT |
L312L |
NP_001145.1 |
View Full Product Details