Product Details

SNP ID
rs1155845
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:210189875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATGGTTGCTATTTAATGGTCAC[C/T]AACAACACATCATTGGCAGACTGGA
Phenotype
MIM: 609576
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ACADL PubMed Links

Gene Details

Gene
ACADL
Gene Name
acyl-CoA dehydrogenase, long chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001608.3 Intron NP_001599.1
XM_005246517.4 Intron XP_005246574.1
XM_017003955.1 Intron XP_016859444.1

View Full Product Details