Product Details

SNP ID
rs816944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64047183 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGGGGTGAGTTAGAACAGCTGA[C/T]GGAGACCCTGGAGTCTGTGTCTGCA
Phenotype
MIM: 601618 MIM: 604784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00176 PubMed Links

Gene Details

Gene
LINC00176
Gene Name
long intergenic non-protein coding RNA 176
There are no transcripts associated with this gene.

Gene
SOX18
Gene Name
SRY-box 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018419.2 Intron NP_060889.1
Gene
TCEA2
Gene Name
transcription elongation factor A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003195.4 Intron NP_003186.1
NM_198723.1 Intron NP_942016.1
XM_005260229.3 Intron XP_005260286.1
XM_011529022.1 Intron XP_011527324.1
XM_011529023.1 Intron XP_011527325.1
XM_011529027.2 Intron XP_011527329.1
XM_017028036.1 Intron XP_016883525.1
XM_017028037.1 Intron XP_016883526.1

View Full Product Details