Product Details

SNP ID
rs816946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64052530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCTTGGCGCGGCTCTGGGCCG[C/G]GATGGCAGGACCTCCCAGCCACTGC
Phenotype
MIM: 601618 MIM: 604784
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SOX18 PubMed Links

Gene Details

Gene
SOX18
Gene Name
SRY-box 18
There are no transcripts associated with this gene.

Gene
TCEA2
Gene Name
transcription elongation factor A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003195.4 Intron NP_003186.1
NM_198723.1 Intron NP_942016.1
XM_005260229.3 Intron XP_005260286.1
XM_011529022.1 Intron XP_011527324.1
XM_011529023.1 Intron XP_011527325.1
XM_011529027.2 Intron XP_011527329.1
XM_017028036.1 Intron XP_016883525.1
XM_017028037.1 Intron XP_016883526.1

View Full Product Details