Product Details

SNP ID
rs926717
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18292955 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGTGGGGAGGAATTGAGAGAGT[C/T]GGCAGAGGTGGAATCCATTTGCCTT
Phenotype
MIM: 604075
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 3138 Intron NP_001076799.1
NM_001282995.1 3138 Intron NP_001269924.1
NM_001282996.1 3138 Intron NP_001269925.1
NM_001282997.1 3138 Intron NP_001269926.1
NM_001282998.1 3138 Intron NP_001269927.1
NM_001282999.1 3138 Intron NP_001269928.1
NM_001283000.1 3138 Intron NP_001269929.1
NM_001283001.1 3138 Intron NP_001269930.1
NM_001283002.1 3138 Intron NP_001269931.1
NM_001283003.1 3138 Intron NP_001269932.1
NM_001283004.1 3138 Intron NP_001269933.1
NM_001283005.1 3138 Intron NP_001269934.1
NM_001283006.1 3138 Intron NP_001269935.1
NM_001283007.1 3138 Intron NP_001269936.1
NM_001283008.1 3138 Intron NP_001269937.1
NM_003434.5 3138 Intron NP_003425.2
XM_005260819.1 3138 Intron XP_005260876.1
XM_005260820.4 3138 Intron XP_005260877.1
XM_011529336.1 3138 Intron XP_011527638.1
XM_011529337.1 3138 Intron XP_011527639.1
XM_011529338.2 3138 Intron XP_011527640.1
XM_011529339.1 3138 Intron XP_011527641.1
XM_011529340.1 3138 Intron XP_011527642.1
XM_017028041.1 3138 UTR 5 XP_016883530.1
XM_017028042.1 3138 Intron XP_016883531.1
XM_017028043.1 3138 UTR 5 XP_016883532.1
XM_017028044.1 3138 UTR 5 XP_016883533.1
XM_017028045.1 3138 UTR 5 XP_016883534.1
XM_017028046.1 3138 UTR 5 XP_016883535.1
XM_017028047.1 3138 Intron XP_016883536.1
XM_017028048.1 3138 Intron XP_016883537.1
XM_017028049.1 3138 UTR 5 XP_016883538.1
XM_017028050.1 3138 Intron XP_016883539.1
XM_017028051.1 3138 Intron XP_016883540.1
XM_017028052.1 3138 Intron XP_016883541.1
XM_017028053.1 3138 Intron XP_016883542.1
XM_017028054.1 3138 Intron XP_016883543.1
XM_017028055.1 3138 Intron XP_016883544.1
XM_017028056.1 3138 Intron XP_016883545.1
XM_017028057.1 3138 Intron XP_016883546.1
XM_017028058.1 3138 Intron XP_016883547.1

View Full Product Details