Product Details

SNP ID
rs926718
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:18292908 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGACTGGCAGGAGGCTTTTGCCATT[A/G]TCCTGGTGCTCCAACTGGTGTAGGG
Phenotype
MIM: 604075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 3091 Intron NP_001076799.1
NM_001282995.1 3091 Intron NP_001269924.1
NM_001282996.1 3091 Intron NP_001269925.1
NM_001282997.1 3091 Intron NP_001269926.1
NM_001282998.1 3091 Intron NP_001269927.1
NM_001282999.1 3091 Intron NP_001269928.1
NM_001283000.1 3091 Intron NP_001269929.1
NM_001283001.1 3091 Intron NP_001269930.1
NM_001283002.1 3091 Intron NP_001269931.1
NM_001283003.1 3091 Intron NP_001269932.1
NM_001283004.1 3091 Intron NP_001269933.1
NM_001283005.1 3091 Intron NP_001269934.1
NM_001283006.1 3091 Intron NP_001269935.1
NM_001283007.1 3091 Intron NP_001269936.1
NM_001283008.1 3091 Intron NP_001269937.1
NM_003434.5 3091 Intron NP_003425.2
XM_005260819.1 3091 Intron XP_005260876.1
XM_005260820.4 3091 Intron XP_005260877.1
XM_011529336.1 3091 Intron XP_011527638.1
XM_011529337.1 3091 Intron XP_011527639.1
XM_011529338.2 3091 Intron XP_011527640.1
XM_011529339.1 3091 Intron XP_011527641.1
XM_011529340.1 3091 Intron XP_011527642.1
XM_017028041.1 3091 UTR 5 XP_016883530.1
XM_017028042.1 3091 Intron XP_016883531.1
XM_017028043.1 3091 UTR 5 XP_016883532.1
XM_017028044.1 3091 UTR 5 XP_016883533.1
XM_017028045.1 3091 UTR 5 XP_016883534.1
XM_017028046.1 3091 UTR 5 XP_016883535.1
XM_017028047.1 3091 Intron XP_016883536.1
XM_017028048.1 3091 Intron XP_016883537.1
XM_017028049.1 3091 UTR 5 XP_016883538.1
XM_017028050.1 3091 Intron XP_016883539.1
XM_017028051.1 3091 Intron XP_016883540.1
XM_017028052.1 3091 Intron XP_016883541.1
XM_017028053.1 3091 Intron XP_016883542.1
XM_017028054.1 3091 Intron XP_016883543.1
XM_017028055.1 3091 Intron XP_016883544.1
XM_017028056.1 3091 Intron XP_016883545.1
XM_017028057.1 3091 Intron XP_016883546.1
XM_017028058.1 3091 Intron XP_016883547.1

View Full Product Details