Product Details
- SNP ID
-
rs816939
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:64037492 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAAGAGCCTGCAGACCAGCCCCACT[A/C]GAGCTGTACGGGGCTGGTGGGGGGC
- Phenotype
-
MIM: 613979
MIM: 604784
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00176
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3764736] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00176
- Gene Name
- long intergenic non-protein coding RNA 176
There are no transcripts associated with this gene.
- Gene
- PRPF6
- Gene Name
- pre-mRNA processing factor 6
There are no transcripts associated with this gene.
- Gene
- TCEA2
- Gene Name
- transcription elongation factor A2
There are no transcripts associated with this gene.
View Full Product Details