Product Details
- SNP ID
-
rs1685312
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219435195 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGTGAGCGGGACGCCCCAGCCCA[G/T]CCTCCGCTGGTTCCGGGATGGGCAG
- Phenotype
-
MIM: 125660
MIM: 615950
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DES
PubMed Links
Gene Details
- Gene
- DES
- Gene Name
- desmin
There are no transcripts associated with this gene.
- Gene
- SPEG
- Gene Name
- SPEG complex locus
View Full Product Details