Product Details

SNP ID
rs1000935
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218826772 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGGCACAGAGAGGCTAAGCAACT[G/T]GCCCAAGTCTCAGAGTAGACGGAGA
Phenotype
MIM: 604976
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MIR9500 PubMed Links

Gene Details

Gene
MIR9500
Gene Name
microRNA 9500
There are no transcripts associated with this gene.

Gene
PRKAG3
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017431.2 Intron NP_059127.2
XM_017004343.1 Intron XP_016859832.1

View Full Product Details