Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001105537.2 | Intron | NP_001099007.1 | |||
XM_006712730.3 | Intron | XP_006712793.1 | |||
XM_011511784.2 | Intron | XP_011510086.1 | |||
XM_011511785.2 | Intron | XP_011510087.1 | |||
XM_011511786.2 | Intron | XP_011510088.1 | |||
XM_011511787.2 | Intron | XP_011510089.1 | |||
XM_011511788.2 | Intron | XP_011510090.1 | |||
XM_011511789.2 | Intron | XP_011510091.1 | |||
XM_017004871.1 | Intron | XP_016860360.1 | |||
XM_017004872.1 | Intron | XP_016860361.1 |