Product Details

SNP ID
rs1803688
Assay Type
DME
NCBI dbSNP Submissions
2
Location
Chr.1:109737041 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CTGACCCCTTACGGACAGGATGAAA[A/C]AAAACAAGCTCTGCAAGTCTGCCTC
Phenotype
MIM: 138390
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GSTM3 PubMed Links

Gene Details

Gene
GSTM3
Gene Name
glutathione S-transferase mu 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000849.4 1018 UTR 3 NP_000840.2

View Full Product Details