Product Details

SNP ID
rs1051950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35498033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGACTACTGCTCCCGGACAGGAA[G/T]TGACTGGGGCGCGGGGAGGAGGGGG
Phenotype
MIM: 603170 MIM: 602280
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TEAD3 PubMed Links

Gene Details

Gene
TEAD3
Gene Name
TEA domain transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003214.3 1967 Intron NP_003205.2
Gene
TULP1
Gene Name
tubby like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289395.1 1967 UTR 3 NP_001276324.1
NM_003322.4 1967 UTR 3 NP_003313.3

View Full Product Details