Product Details

SNP ID
rs274305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:24958051 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTGTAATTGAAGGAACAGTTCA[A/G]TCTATCCACTGAATTGTTCCTAGCC
Phenotype
MIM: 610751 MIM: 611260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRTFDC1 PubMed Links

Gene Details

Gene
PRTFDC1
Gene Name
phosphoribosyl transferase domain containing 1
There are no transcripts associated with this gene.

Gene
THNSL1
Gene Name
threonine synthase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024838.4 Intron NP_079114.3
XM_005252597.2 Intron XP_005252654.1
XM_017016665.1 Intron XP_016872154.1

View Full Product Details