Product Details

SNP ID
rs784368
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:211383328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTCCAACTTTCTTTGTAATTATG[C/T]CCTCTGTTTCAGAATCCTACTGCCG
Phenotype
MIM: 602356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

Gene
TRAF5
Gene Name
TNF receptor associated factor 5
There are no transcripts associated with this gene.

View Full Product Details