Product Details

SNP ID
rs784377
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:211392298 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATGGGGACAAGGAAGGGCTTTG[A/G]AGAGGAAATGATTATCTAAACTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

View Full Product Details